Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-Well Filter Plates with Ion Exchange Membrane
Optimized for a variety of applications requiring larger per well volume processing.
Brand:  Cytiva (formerly Pall Lab) 8171
Additional Details : Weight : 1.25400kg
 View more versions of this product
Product Code. 17124631
  • £233.00 / Pack of 5
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
Description
Description
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Mustang Q (Anion Exchange)
Non-sterile
1 mL
0.8 μm
5/Pk.