Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Multiplex Assays
Optimized for a variety of applications requiring larger per well volume processing.
Brand:  Cytiva (formerly Pall Lab) 8049
Additional Details : Weight : 0.92000kg
Product Code. 17154611
  • £233.00 / Pack of 10
Estimated Shipment: 22-07-2024
to see stock.
Add to basket
Description
Description
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Polypropylene/Polyethylene Non-Woven and Supor (Polyethersulfone)
1.2 μm
Non-sterile
10/Pk.