Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Ultrafiltration
Optimized for a variety of applications requiring larger per well volume processing.
Brand:  Cytiva (formerly Pall Lab) 8034
Additional Details : Weight : 0.85500kg
 View more versions of this product
Product Code. 17184601
  • £677.00 / Pack of 10
Estimated Shipment: 05-09-2024
to see stock.
Add to basket
Description
Description
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Omega (Modified PES)
Non-sterile
10/Pk.
10 kDa
Polypropylene Housing
350 μL