missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Glass Wool
Filter aid
£23.62 - £89.14
Specifications
Boiling Point | 1000°C |
---|---|
Product Type | Glass Wool |
For Use With (Application) | Filter aid |
Material | Glass |
Melting Point | 680°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10637274
|
Thermo Scientific Chemicals
386060500 |
50g |
£23.62
50g |
In Stock
Log in to see stock. |
|||||
10024923
|
Thermo Scientific Chemicals
386062500 |
250g |
£31.96
250g |
In Stock
Log in to see stock. |
|||||
11927001
|
Thermo Scientific Chemicals
386060010 |
1kg |
£89.14
1kg |
In Stock
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1000°C | |
Filter aid | |
680°C |
Glass Wool | |
Glass |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Glass wool
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title