missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å Product Code.: 15678625

Thermo Scientific Chemicals Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å

Product Code. 15678625
5 kg
Click to view available options
Quantity:
1 kg
5 kg
30 kg
This item is not returnable. View return policy

Product Code. 15678625

Brand: Thermo Scientific Chemicals 240370050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For column chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9

Specifications

Packaging Plastic Drum
Quantity 5 kg
Type Silica Gel
Material Silica Gel
For Use With (Application) Column Chromatography

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466634 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog Number240370050,240370300
Lot NumberA0459322 Certificate TypeCertificate of Analysis Date15/01/2024 Catalog Number240370050,240370300
Lot NumberA0452105 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number240370050,240370300
Lot NumberA0411572 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number240370050,240370010,240370300
Lot NumberA0417464 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number240370050,240370010,240370300

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.