missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide, >99%, Molecular biology reagent grade, MP Biomedicals™
Click to view available options
Quantity:
500 g
Specifications
CAS | 79-06-1 |
Color | White |
Density | 1.127g/cm3 at 25°C |
Molecular Formula | C3H5NO |
MDL Number | MFCD00008032 |
Quantity | 500 g |
Synonym | acrylamide, 2-propenamide, propenamide, acrylic amide, akrylamid, ethylenecarboxamide, vinyl amide, acrylic acid amide, 2-propeneamide, propeneamide |
InChI Key | HRPVXLWXLXDGHG-UHFFFAOYSA-N |
SMILES | NC(=O)C=C |
IUPAC Name | prop-2-enamide |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'