missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide-bis-acryl, 40% (w/v), 29:1 Solution, MP Biomedicals™
Acrylamide-bis-acryl, 40% (w/v), 29:1 Solution is sequencing gel that provides convenient ready-to-use solutions of 4X high quality acrylamide for molecular biology.
Brand: MP Biomedicals 11BIAC2902
This item is not returnable.
View return policy
Description
- Acrylamide/bis-acrylamide (40% w/v) 29:1 Solution is convenient ready-to-use solutions of 4X crystalized high quality acrylamide for molecular biology, and NN'-Methylenebisacrylamide in deionised water.
- Premixed 19:1 liquid solution eliminates the need to weigh toxic acrylamide and bis-acrylamide.
- The concentration is based on the total weight of both the acrylamide and bis-acrylamide.
- Acrylamide/bis-acrylamide (40% w/v) 29:1 Solution is prepared from highly purified acrylamide (4X) and bis-acrylamide (2X) using deionized ultrafiltered water.
Specifications
A290 (1% in water): ≤0.1 | |
Acrylamide/Bis-acrylamide 29:1 | |
Molecular Biology |
Liquid | |
1 L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- acrylamide/bis-acrylamide 29:1
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H372-Causes damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acrylamide, N,N'-methyl-bis-acrylamide
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.