missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide: Bis-Acrylamide 29:1 (40% Solution/Electrophoresis), Fisher BioReagents
Brand: Fisher BioReagents BP1408-1
90.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
PP Bottle; 1 L
This Acrylamide Solution is premixed for 3.3% cross-linking and is suitable for DNA sequencing and separation of proteins.
Specifications
Acrylamide: Bis-Acrylamide 29:1 | |
Liquid | |
10μmhos max. | |
Colorless | |
Polypropylene Bottle | |
C3H5NO | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
prop-2-enamide | |
6579 | |
Electrophoresis |
40% Solution | |
79-06-1, 110-26-9, 7732-18-5 | |
1.035 g/mL | |
1 L | |
DNase-, RNase- and Protease-Free | |
MFCD00008032 | |
NC(=O)C=C | |
71.08 | |
CHEBI:28619 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Acrylamide : Bis-Acrylamide 29:1 (40% Solution/Electrophoresis)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H372-Causes damage to organs.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H361f-Suspected of damaging fertility.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Supplemental information
- MIXTURE LIST-Contains : acrylamide, methylene diacrylamide
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Acrylamide: Bis-Acrylamide 29:1 (40% Solution/Electrophoresis), Fisher BioReagents > PP Bottle; 1 L
Spot an opportunity for improvement?Share a Content Correction