missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide/Bis Solution, 29:1, (30% w/v), 3.3% C, Serva Electrophoresis™
Specifications
Physical Form | Liquid |
---|---|
Composition | Acrylamide/Bis Solution, 29:1 |
Conductivity | <100μS/cm |
Density | 1.04 at 20°Cg/cm3 |
Color | Colorless |
pH | 6 to 8 |
Solubility Information | Fully miscible |
Odor | Odorless |
Specifications
Liquid | |
<100μS/cm | |
Colorless | |
Fully miscible |
Acrylamide/Bis Solution, 29:1 | |
1.04 at 20°Cg/cm3 | |
6 to 8 | |
Odorless |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acrylamide/Bis Solution, 29:1
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1
- Carcinogenicity Category 1
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.