Learn More
Thermo Scientific Chemicals Acrylamide/Bisacrylamide 29:1, powder
Premixed powder for preparing protein DNA gels
Brand: Thermo Scientific Chemicals J60824.22
64.98 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.10000kg Transport : UN number : 2074 Chem class : 6.1 Pack group : III
Description
Acrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAcrylamide/Bisacrylamide 29:1, Premixed powder is used for preparing protein DNA gels. It is used as a DNA sequencing gels, Protein separation. They are also used for preparing gels using stock solutions with the most common cross-link ratios.
Solubility
It is soluble in water.
Notes
Keep away from light. Store at room tempeature.
Specifications
Acrylamide/Bisacrylamide 29:1 | |
White | |
MFCD00080848 | |
It is soluble in water. |
Powder | |
100 g | |
UN2074 |
RUO – Research Use Only