missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Acrylamide / N,N'-Methylenebisacrylamide 19:1, for biochemistry, 40% mix solution in water
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
Specifications
Specifications
| Absorbance | (1% in water), (1% in water) at 0nm, 0.1 max. at 290nm |
| Chemical Name or Material | Acrylamide / N, N'-Methylenebisacrylamide 19:1 |
| Physical Form | Liquid |
| Acrylic Acid | 0.001% max. |
| CAS | 79-06-1, 7732-18-5, 110-26-9, 79-06-1 |
| CAS Min % | 59.0 |
| CAS Max % | 61.0 |
| Conductivity | 3μS/cm Max. |
| Color | Colorless |
| Quantity | 500 mL |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction