Learn More
Thermo Scientific Chemicals Acrylamide / N,N'-Methylenebisacrylamide 37.5:1, for biochemistry, 40% mix solution
Acrylamide / N,N'-Methylenebisacrylamide, CAS # 79-06-1, 7732-18-5, 110-26-9, 79-06-1, is a water soluble compound that cross-links to produce polyacrylamide gels that are widely used in chromatography and the electrophoretic separation of nucleic acids and proteins.
Brand: Thermo Scientific Chemicals 330221000
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
- Acrylamide / N,N'-Methylenebisacrylamide is a water-soluble, odorless, white crystalline powder
- This compound polymerizes in the presence of free radicals to generate gels whose pore size may be regulated by manipulating the relative concentrations of this mixture of compounds
Applications
- Polyacrylamide gels are widely used in biochemistry, forensic chemistry, genetics, molecular biology, and biotechnology to separate proteins or nucleic acids, according to their electrophoretic mobility
- Because of their higher resolution compared to other gels, polyacrylamide gels are also used in the analysis of smaller RNA species from mixed samples
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
(1% in water), (1% in water) at 0nm, 0.1 max. at 290nm | |
Liquid | |
79-06-1, 7732-18-5, 110-26-9, 79-06-1 | |
61.0 | |
Colorless | |
Glass Bottle | |
MFCD00008032 | |
NC(=O)C=C | |
71.08 | |
CHEBI:28619 |
Acrylamide / N, N'-Methylenebisacrylamide 37.5:1 | |
0.001% max. | |
59.0 | |
3μS/cm Max. | |
100 mL | |
C3H5NO | |
HRPVXLWXLXDGHG-UHFFFAOYSA-N | |
prop-2-enamide | |
6579 | |
Biochemical |
Safety and Handling
- Acrylamide / N,N``-Methylenebisacrylamide 37.5:1, 40% mix solution inwater
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Acrylamide, Methylene diacrylamide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only