missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide (White, Needle-like Crystals/Electrophoresis), Fisher BioReagents
96.73 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
5 kg
Packaging:
Poly Bottle
Poly Pail
Description
This Acrylamide is 3X recrystallized to obtain a high-purity crystalline material suitable for electrophoretic separation of proteins and nucleic acids.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Solid |
Acrylic Acid | 0.002% max. |
Boiling Point | 125°C |
Melting Point | 82°C |
Conductivity | 2.50μmhos max. |
Color | White |
pH | 6.5 to 8.0 |
Quantity | 100 g |
Assay Percent Range | ≥99.9 % |
DNase | DNase free |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- Acrylamide
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- H317-May cause an allergic skin reaction.
- H372-Causes damage to organs.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H361f-Suspected of damaging fertility.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H301-Toxic if swallowed.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'