missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Acrylic Safety Glasses Dispenser
Access safety accessories with ease. Fisherbrand™ Acrylic Safety Glasses Dispenser holds protective goggles, other safety items and personal items to keep them safe and clean. They are ergonomically angled for easy retrieval. Transparent blue dispensers are available in two sizes.
£145.00 - £249.00
Specifications
Color | Blue |
---|---|
Product Type | Dispenser |
For Use With (Equipment) | Safety Glasses |
Includes | Wall mounting screws |
No. per Pack | 1 Ea. |
Description
- Translucent acrylic dispensers provide easy access to safety accessories
- Ergonomically angled for easy dispensing
- Hinged lid protects glasses from dust and debris
- Flip open top for filling with tilt-open bottom for dispensing
- Set on counter or wall mount (screws included) for convenient access
- Large dispenser holds approximately 20 pairs of glasses
Small dispenser holds approximately 10 pairs of glasses
Specifications
Blue | |
Safety Glasses | |
1 Ea. |
Dispenser | |
Wall mounting screws | |
Acrylic |