missing translation for 'onlineSavingsMsg'
Learn More
Megro™ Acrylic Glass Glove Box Holder Product Code.: 15752728
Add to basket

Megro™ Acrylic Glass Glove Box Holder

Product Code. 15752728
Glove box holder; acrylic plexiglass
Click to view available options
Glove box holder; acrylic plexiglass

Product Code. 15752728

Brand: Megro™ MEG136601

Add to basket
Add to basket

For ready-to-hand storage of examination gloves.

  • Includes wall mounting material
  • 25 x 13 x 7.5cm
  • Transparent for easy view of contents
  • Acrylic plexiglass

Specifications

Color Clear
Dimensions (L x W x H) 25 x 13 x 7.5 cm
Holds Gloves
Material Acrylic Plexiglass
Mounting Wall Mount
For Use With (Application) Examination glove box storage
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Megro™ Acrylic Glass Glove Box Holder >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.