missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Acrylic 1-Compartment Dispensing Bin

Keep spare lab and office items easily accessible. Fisherbrand™ Acrylic Dispensing Bins have large openings with hinged lids for easy filling and dispensing. The sturdy clear bins make it easy to monitor inventory levels.

Brand:  Fisherbrand™ HS23401F

Product Code. 11764834

  • £138.32 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Ideal for pipette tips, tubes, fittings, syringe filters, septa and other small lab and office items
  • Made from 3mm thick, transparent acrylic for at-a-glance inventory monitoring
  • Hinged lids over generous-sized openings for rapid filling and dispensing
  • Flip-top cover keeps products secure and dust free
  • Includes non-skid rubber feet
Specifications

Specifications

Dispensing Bin
Transparent
194 mm
184 mm
Acrylic
1/Pk.
194 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Acrylic 1-Compartment Dispensing Bin > Depth: 194mm; Dimensions: 184W x 232mmH

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.