missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Deutsch Neumann™ Acrylnitril-Butadien Stoppers
£0.55 - £1.95
Product Code | Brand | Diameter (Metric) Bottom | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Bottom | Price | Quantity & Availability | |||||
10411104
|
Deutsch Neumann™
1010323 |
23 mm |
£1.00
Each |
In Stock
Log in to see stock. |
|||||
10340671
|
Deutsch Neumann™
1010321 |
21 mm |
£0.75
Each |
In Stock
Log in to see stock. |
|||||
10579412
|
Deutsch Neumann™
1010329 |
29 mm |
£1.45
Each |
In Stock
Log in to see stock. |
|||||
10014770
|
Deutsch Neumann™
1010318 |
18 mm |
£0.55
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
10606371
|
Deutsch Neumann™
1010331 |
31 mm |
£1.95
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||||
Specifications
Black | |
45 Shore A |