missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSF, Tocris Bioscience™
Click to view available options
Quantity:
25 mL
Description
Artificial cerebrospinal fluid (aCSF) commonly used when sampling from brain interstitial fluid. Closely matches the electrolyte concentrations of CSF and is prepared from high purity water and analytical grade reagents. Microfiltered and sterile. Final ion concentrations (in mM): Na+ 150; K+ 3.0; Ca2+ 1.4; Mg2+ 0.8; P 1.0; Cl- 155.
Specifications
Quantity | 25 mL |
Synonym | Artificial cerebrospinal fluid |
Percent Purity | >98% |
Chemical Name or Material | ACSF |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'