missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20710-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to ACSL3.
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme is highly expressed in brain, and preferentially utilizes myristate, arachidonate, and eicosapentaenoate as substrates. The amino acid sequence of this isozyme is 92% identical to that of rat homolog. Two transcript variants encoding the same protein have been found for this gene.Specifications
ACSL3 | |
Polyclonal | |
Unconjugated | |
ACSL3 | |
ACS3, ACSL3, FACL3, LACS 3, LACS3, PRO2194 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2181, 74205 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95573, Q9CZW4 | |
Acsl3 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |