Learn More
Tocris Bioscience™ ACT 462206
Potent, dual orexin receptor antagonist
Brand:  Tocris Bioscience™ 5319/10
164.14 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18770528
  • £206.00 / 10mg
Explore more special offers
Description
Description
ACT 462206 is a potent, dual orexin receptor antagonist (IC50 values are 11 and 60 nM for OX2 and OX1 receptors respectively). Exhibits anxiolytic-like properties in vivo. Brain penetrant.
Specifications
Specifications
>99%
ACT 462206
ACT 462206
10 mg
388.48