Learn More
Invitrogen™ Actinomycin D
Brand: Invitrogen™ 11805017
Code : 50
Additional Details : CAS Number : 50-76-0 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : II
Description
- Acts by interfering with mRNA synthesis
- ctinomycin D is used as a selective antibiotic for resistant bacteria, generally at a concentration of about 1μg/mL
- Product is provided as a powder
- Made into a stock solution at 1mg/ml in DMSO
Cell Analysis, Cell Structure, Cloning, Nucleus, Nucleoli & Nuclear Envelope, Transformation
Order Info
Shipping Condition: Room Temperature
Compliance
Manufactured at facilities that comply with cGMP manufacturing requirements and that are certified to ISO 13485 and registered with FDA as medical device manufacturers.
Specifications
1 μg/mL | |
Powder | |
Antibiotic |
Safety and Handling
- Actinomycin D
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Carcinogenicity Category 1A
- Reproductive toxicity Category 1A
Hazard Statement
- H300-Fatal if swallowed.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P363-Wash contaminated clothing before reuse.
Supplemental information
- MIXTURE LIST-Contains : Actinomycin D
For Research Use Only. Not for use in diagnostic procedures.