missing translation for 'onlineSavingsMsg'
Learn More

Actinomycin D (Red Crystals), Fisher BioReagents

Product Code. 10688203
Click to view available options
Quantity:
5 mg
10 mg
Packaging:
Poly Micro Tube
This item is not returnable. View return policy
This item is not returnable. View return policy

Isolated from Streptomyces parvuius

This antibiotic is a widely used transcription inhibitor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-76-0
Molecular Formula C62H86N12O16
Molecular Weight (g/mol) 1255.438
InChI Key RJURFGZVJUQBHK-HUZQGMAJSA-N
PubChem CID 131954673
IUPAC Name 2-amino-4,6-dimethyl-3-oxo-1-N-[(3R,6S,7R,10S,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,11,14-tetrazabicyclo[14.3.0]nonadecan-6-yl]-9-N-[(3R,6S,7R,16S)-7,11,14-trimethyl-2,5,9,12,15-pentaoxo-3,10-di(propan-2-yl)-8-oxa-1,4,
SMILES CC1C(C(=O)NC(C(=O)N2CCCC2C(=O)N(CC(=O)N(C(C(=O)O1)C(C)C)C)C)C(C)C)NC(=O)C3=C4C(=C(C=C3)C)OC5=C(C(=O)C(=C(C5=N4)C(=O)NC6C(OC(=O)C(N(C(=O)CN(C(=O)C7CCCN7C(=O)C(NC6=O)C(C)C)C)C)C(C)C)C)N)C

Specifications

Type Actinomycin D
Physical Form Powder/Solid
Melting Point 245°C
Extinction Coefficient Report
Assay Percent Range ≥98 %
Merck Index 15, 2797
Packaging Poly Micro Tube
Solubility Information Soluble in Methanol
Formula Weight 1255.44
Percent Purity 99.0% min. (HPLC), 98% min. (TLC)
Color Red
Quantity 5 mg
Chemical Name or Material Actinomycin-D
Show More Show Less
compliance-icons
Product Identifier
  • Actinomycin D
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'