missing translation for 'onlineSavingsMsg'
Learn More

Cytiva ActiSM liquid medium

A chemically defined ADCF medium optimized for high-yield protein production. Designed for use with ActiPro medium.

Brand:  Cytiva SH31040.02

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15842962

  • £89.11 / 1000mL
Estimated Shipment: 28-08-2024
to see stock.

Description

Description

  • HyClone ActiSM cell culture medium has been formulated to provide high yields of recombinant proteins in cell culture processes when using Chinese hamster ovary (CHO) cell lines.
  • The medium is chemically defined, animal-derived component-free (ADCF), and optimized for high-yield protein production.
  • ActiSM medium does not contain any growth factors (such as insulin), peptides, hydrolysates, phenol red, or 2-mercaptoethanol, ensuring batch-to-batc
  • ActiSM medium is intended for use in combination with HyClone ActiPro medium and Cell Boost™ 7a and Cell Boost 7b supplements as part of the ActiPro system of media and supplements designed with adaptation and production in mind.
  • The ActiPro system is suitable for general biomanufacturing with CHO cell lines such as CHO-GS, CHO-K1, CHO-DG44, and CHO-S.
Specifications

Specifications

2 to 8 °C, away from light
Sterile
Poloxamer 188
Bottle
Liquid
Sterile-filtered
No Glutamine
1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers