Learn More
Ansell ActivArmr™ 97-013 Medium Duty Hi-Viz™ Gloves with Intercept™ Technology Product Code.: 16895260
Add to basket
Ansell ActivArmr™ 97-013 Medium Duty Hi-Viz™ Gloves with Intercept™ Technology
Product Code. 16895260 Shop All Ansell Products
10
Click to view available options
Size:
7
8
9
10
11
Product Code. 16895260
Brand: Ansell 97013100
Add to basket
Add to basket
Ergonomically designed gloves reduce hand fatigue, while highly visible coloring on the back of the hand and palm side increases safety in low light conditions.
  • Highly reflective back keeps workers safe in low-visibility and dark environments
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • 13-gauge liner offers increases comfort for all day wear
  • Breathable fabric minimizes perspiration compared to leather
  • Ergonomic design provides naturally curved, relaxed fit to reduce hand fatigue
  • Enhances grip capabilities in wet and dry conditions
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color Hi-vis Orange
Cut Resistance Rating ANSI A2
Hand Specific
Material Intercept Technology Yarn
Size 10
EN388 Standard 4X32B
Glove Cuff Style Knit
Series 97-013
Yarn Gauge 13
Product Title
Ansell ActivArmr™ 97-013 Medium Duty Hi-Viz™ Gloves with Intercept™ Technology >