missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ Winter Monkey Grip™ 23-173 Heavy Duty PVC-Coated Insulated Gloves

Product Code. 12650782
Click to view available options
Size:
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Textured PVC coating protects hands from abrasions and provides water, oil, and grease repellency. Foam insulation remains flexible, and helps to keep hands warm in cold applications.

  • Textured PVC vinyl coating protects hands from abrasions and provides water, oil, and grease repellency
  • Coating remains soft and flexible in cold conditions
  • Foam insulation helps to keep hands warm
  • Enhanced grip capabilities when handling heavy-duty objects
  • Curved fingers and wing thumb increase user comfort
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Coating Material PVC
Color Orange
Cut Resistance Rating ANSI A1
Exterior Finish Fully Textured
Hand Specific
Size 11
EN388 Standard 3241B
For Use With (Application) Cold Storage, Equipment Repair, Metal Fabrication, and Warehousing
Glove Cuff Style Safety
Liner Material Cotton
Series 23-173
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'