missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated charcoal, NORIT(R) A SUPRA, Thermo Scientific Chemicals
Activated charcoal,NORIT™ A SUPRA, C, CAS Number-7440-44-0, acticarbone, activated charcoal, anthrasorb, carbon-12, carbono, carbosieve, graphene, graphite, mineral, norit, 1kg, Passes Test, CHEBI:27594, Black, 231-153-3, OKTJSMMVPCPJKN-UHFFFAOYSA-N, 15% max., 3652.0 deg.C, 15, 1807
£57.18 - £197.99
Chemical Identifiers
CAS | 7440-44-0 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12 |
MDL Number | MFCD00133992 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10419640
|
Thermo Scientific Acros
134342500 |
250 g | Glass bottle |
£57.18
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10289873
|
Thermo Scientific Acros
134340010 |
1 kg | Plastic drum |
£197.99
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aidChemical Identifiers
7440-44-0 | |
12 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Specifications
Passes Test | |
3652.0°C | |
15% max. | |
MFCD00133992 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
12 | |
Activated charcoal,NORIT* A SUPRA |
7440-44-0 | |
Black | |
C | |
15, 1807 | |
Solubility in water: insoluble. Other solubilities: insoluble in organic solvents,soluble matter in hno3: <=3%,soluble matter in alcohol: <=0.5% | |
[C] | |
12 | |
CHEBI:27594 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 231-153-3
missing translation for 'rtecsNumber' : FF5250100
missing translation for 'tsca' : TSCA
Product Identifier
- Activated charcoal
RUO – Research Use Only