missing translation for 'onlineSavingsMsg'
Learn More

Activated charcoal, NORIT(R) GAC 1240, 12-40 mesh, Thermo Scientific Chemicals

£46.23 - £110.55

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10463935
missing translation for 'viewDocuments'
Thermo Scientific Acros
395712500
250 g Glass bottle
£46.23
250g
Please to purchase this item. Need a web account? Register with us today!
10269043
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific Acros
395710010
1 kg Glass bottle
£110.55
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
Specifications

Chemical Identifiers

7440-44-0
12
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00133992
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]

Specifications

7440-44-0
Black
12 mesh (min. 5%), 40 mesh (max. 4% max.)
MFCD00133992
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
12
Activated charcoal,NORIT* GAC 124
3652.0°C
2% max. (as packed)
C
15, 1807
Solubility in water: insoluble. Other solubilities: insoluble in organic solvents,soluble matter in hno3: <=3%,soluble matter in alcohol: <=0.5%
[C]
12
CHEBI:27594
Granular Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-153-3

Product Identifier
  • Activated charcoal

SDS
Documents

Documents

Certificates

RUO – Research Use Only