missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal, Tested according to Ph.Eur., Honeywell™ Fluka™
Click to view available options
Quantity:
1 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | 3550°C (literature) |
Color | Black |
Density | ca. 350 kg/cu.m |
pH | 6 to 9 |
Packaging | Plastic Bottle |
Quantity | 1 kg |
UN Number | NONH for all modes of transport |
Formula Weight | 12.01g/mol |
Chemical Name or Material | Activated Charcoal, Tested According to Ph.Eur. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'