missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activated Charcoal, Untreated, 4 - 8 Mesh, Honeywell Fluka™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Black |
pH | 6 - 9 |
Mesh Size | 4 to 8mesh |
Odor | Odorless |
Packaging | Plastic bottle |
Quantity | 500 g |
UN Number | NONH for all modes of transport |
Formula Weight | 12.01g/mol |
Physical Form | Powder |
Chemical Name or Material | Activated charcoal |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'