missing translation for 'onlineSavingsMsg'
Learn More

Activated Charcoal, Untreated, 8 - 20 Mesh, Honeywell Fluka™

Product Code. 15637740
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Drum with liner
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Untreated, granular, 8-20 mesh

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Black
pH 6-9
Mesh Size 8 to 20mesh
Odor Odorless
Packaging Poly bottle
Quantity 500 g
UN Number NONH for all modes of transport
Formula Weight 12.01g/mol
Physical Form Powder
Chemical Name or Material Activated charcoal
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'