Learn More
FTM Technologies™ Active Carbon Filters
Used to eliminate noxious or unpleasant smells emanating from liquids, vapours or smoke. FTM Technologies™ Active Carbon Filters are for use with fume hoods.
Brand:  FTM Technologies™ FCP1
Additional Details : Weight : 2.52000kg
 View more versions of this product
Product Code. 11738044
  • £287.77 / Each
In Stock
to see stock.
Description
Description
  • Superior protection in the laboratory
  • Compatible with a wide range of fume hoods
Specifications
Specifications
Fume Hoods (Except for HI8F/HI8V)
For Organic Vapors, Acids and Odors
Filter
Activated Carbon
1Each