Learn More
Socorex™ Acura™ Electro 956.12 Micropipette
Lightweight and ergonomically friendly, this electronic multi-channel micropipette comes with a QC certificate, Qualitips™ pipette tip samples and operating instructions. Socorex™ Acura™ Electro 956.12 Micropipette charges quickly and has a long working autonomy.
Brand: Socorex™ 956.12.350
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Exceptional, user-friendly ergonomics.
- Lightweight.
- Easy, intuitive, self-teaching program.
- Large display, reversible right/left reading.
- Microprocessor-controlled motor.
- Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
- Sequential tip ejection.
- Also fit any single channel volumetric module.
Specifications
<±1.0% - <±0.6% | |
EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) | |
12 | |
Electronic Micropipette | |
Right/Left LCD | |
5.0μL | |
1 Each | |
24 Months |
4.8V NiMH | |
Acura™ 956 | |
Acura™ | |
20 to 350 μL | |
QC certificate, Qualitips™ pipette tip samples and operating instructions. | |
<0.6% - <0.25% | |
<1.5Hours |