missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Acura™ Electro 956.12 Micropipette Initial Kit
Ergonomic electronic multi-channel micropipette initial package. Socorex™ Acura™ Electro 956.12 Micropipette Initial Kit includes pipette, charging stand, fast charger with AC adapter and cord, additional battery pack, and pipette tip samples.
£1235.00 - £1275.00
Specifications
Battery Type | 4.8V NiMH |
---|---|
Certifications/Compliance | EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) |
Model | Acura™ 956 |
Number of Channels | 12 |
Product Line | Acura™ |
Product Code | Brand | Accuracy | Increments | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Accuracy | Increments | Volume (Metric) | Price | Quantity & Availability | |||
11899233
|
Socorex™
956.12.010E |
<±3.5% - <±1.0% | 0.05μL | 0.5 to 10 μL |
£1275.00
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||
11819243
|
Socorex™
956.12.050E |
<±1.0% - <±0.8% | 0.25μL | 2.5 to 50 μL |
£1235.00
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||
Description
- Exceptional, user-friendly ergonomics.
- Lightweight.
- Easy, intuitive, self-teaching program.
- Large display, reversible right/left reading.
- Microprocessor-controlled motor.
- Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
- Sequential tip ejection.
- Also fit any single channel volumetric module.
Specifications
4.8V NiMH | |
Acura™ 956 | |
Acura™ | |
Right/Left LCD | |
1 Each | |
24 Months |
EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS) | |
12 | |
Electronic Micropipette Kit | |
Pipette charging stand, fast charger with AC adapter and cord, additional battery pack, QC certificate, Qualitips™ pipette tip samples and operating instructions. | |
<1.5Hours |