Learn More
Socorex™ Acura™ Manual 815 Fixed Volume Micropipettes
Slim and precise micropipettes set at a fixed volume for easy dose repetition. Socorex™ Acura™ Manual 815 Fixed Volume Micropipettes are easy to handle and have a soft plunger stroke.
Brand:  Socorex™ 815.0500
 View more versions of this product
Product Code. 10365312
  • £177.00 / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Air-displacement fixed-volume pipette.
    • Exceptional, user-friendly ergonomics.
    • Justip™ adjustable tip ejector fitting most tips.
    • Shock, UV-light and autoclaving resistance.
    • Ultra-soft plunger stroke.
    • Available in a variety of volume models.
Specifications
Specifications
<±0.7%
Autoclavable
Polycarbonate
Micropipette
<0.3%
36 Months
ISO 8655, GMP, GLP, NCCLS, CE IVD Directive 98/79 EEC (in vitro diagnostic medical devices).
White
Acura™ Manual 815
500 μL
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products