Learn More
Socorex™ Acura Variable Volume Manual Pipettes
Secure the stability of your settings when pipetting with Socorex™ Acura Variable Volume Manual Pipettes. Each individual unit's accuracy has been measured for reliability at 0.5μL and 1μL.
Brand: Socorex™ 825.0010Y
Code : 04
Additional Details : Weight : 0.16200kg
Description
- Constructed of polypropylene
- With precision setting mechanism
- Single-handed volume setting
Specifications
Autoclavable | |
825 | |
Acura™ | |
1 to 10 μL |
Polypropylene | |
1 | |
Micropipette | |
1 Each |