Learn More
Socorex™ Acura Variable Volume Manual Pipettes
Secure the stability of your settings when pipetting with Socorex™ Acura Variable Volume Manual Pipettes. Each individual unit's accuracy has been measured for reliability at 0.5μL and 1μL.
Brand:  Socorex™ 825.0200
 View more versions of this product
Product Code. 10292102
  • £294.00 / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Constructed of polypropylene
  • With precision setting mechanism
  • Single-handed volume setting
Specifications
Specifications
Autoclavable
825
Acura
20 to 200 μL
Polypropylene
1
Micropipette
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Socorex™ Acura Variable Volume Manual Pipettes > Volume: 20 to 200μL