Learn More
Socorex™ Acura Variable Volume Manual Pipettes
Secure the stability of your settings when pipetting with Socorex™ Acura Variable Volume Manual Pipettes. Each individual unit's accuracy has been measured for reliability at 0.5μL and 1μL.
Brand:  Socorex™ 825.0050
 View more versions of this product
Product Code. 10630663
  • £256.00 / Each
Estimated Shipment: 02-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Constructed of polypropylene
  • With precision setting mechanism
  • Single-handed volume setting
Specifications
Specifications
Autoclavable
825
Acura™
5 to 50 μL
Polypropylene
1
Micropipette
1 Each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products