missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Acura Variable Volume Manual Pipettes
Secure the stability of your settings when pipetting with Socorex™ Acura Variable Volume Manual Pipettes. Each individual unit's accuracy has been measured for reliability at 0.5μL and 1μL.
£245.00 - £289.00
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polypropylene |
Number of Channels | 1 |
Product Line | Acura™ |
Quantity | 1 Each |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10630663
|
Socorex™
825.0050 |
5 to 50 μL |
£245.00
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
|||||
10252012
|
Socorex™
825.0010Y |
1 to 10 μL |
£245.00
Each |
Estimated Shipment: 30-10-2024
Log in to see stock. |
|||||
10393412
|
Socorex™
825.0010 |
0.5 to 10 μL |
£261.00
Each |
Estimated Shipment: 04-11-2024
Log in to see stock. |
|||||
10292102
|
Socorex™
825.0200 |
20 to 200 μL |
£275.54
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
10794913
|
Socorex™
825.1000 |
100 to 1000 μL |
£267.00
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
10698542
|
Socorex™
825.0100 |
10 to 100 μL |
£245.00
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
10445842
|
Socorex™
825.0020 |
2 to 20 μL |
£245.00
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
10531452
|
Socorex™
835.10 |
1 to 10 mL |
£289.00
Each |
Estimated Shipment: 02-09-2024
Log in to see stock. |
|||||
Description
- Constructed of polypropylene
- With precision setting mechanism
- Single-handed volume setting
Specifications
Autoclavable | |
1 | |
1 Each |
Polypropylene | |
Acura™ |