missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ACVR1B (ALK4), GST Tag Recombinant Protein

Human ACVR1B (ALK4), GST Tag Recombinant Protein

Brand:  Invitrogen™ PV4329

Code : NEW

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 10240533

  • £6900.00 / 1mg
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

Description

Description

  • ACVR1B is a serine/threonine kinase involved in signal transduction and cellular communication
  • Several truncated ACVR1B isoforms have been reported to be exclusively expressed in human pituitary tumors
  • Mutations of ACVR1B have been observed in pancreatic adenocarcinomas
Specifications

Specifications

Kinase Assay
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton™ X-100, 2 mM DTT, and 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
1 mg
Baculovirus
GST Tag
Drug Development
Ser/Thr Kinases: TKL Group
68.2 kDa
Recombinant Human Protein
Frozen
Catalytic
Dry Ice
Human
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers