missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ACVR1B (ALK4), GST Tag Recombinant Protein
Human ACVR1B (ALK4), GST Tag Recombinant Protein
Brand: Invitrogen™ PV4312
Code : 50
Additional Details : Weight : 0.05250kg
Description
ACVR1B is a serine⁄threonine kinase involved in signal transduction and cellular communication. Several truncated ACVR1B isoforms have been reported to be exclusively expressed in human pituitary tumors. Mutations of ACVR1B have been observed in pancreatic adenocarcinomas.Specifications
NP_004293 | |
3458, 91 | |
10 μg | |
STKR Kinase Family | |
Human | |
GST Tag | |
Biochemical binding/kinetic, Biochemical phospho-specific activity (no antibody) | |
Dry Ice | |
ACVR1B | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
68.2 kDa | |
ACVR1B (ALK4) | |
aa 150-505 | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton™ X-100, 2 mM DTT, and 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Ser⁄Thr Kinases: TKL Group | |
Z'-LYTE™ | |
activin A receptor, type IB | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |