Learn More
Invitrogen™ Human ACVR1B (ALK4), GST Tag Recombinant Protein
Human ACVR1B (ALK4), GST Tag Recombinant Protein
Brand: Invitrogen™ PR7950A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Several truncated ACVR1B isoforms have been reported to be exclusively expressed in human pituitary tumors. Mutations of ACVR1B have been observed in pancreatic adenocarcinomas.
Specifications
NP_004293 | |
68.2 kDa | |
ACVR1B (ALK4) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: TKL Group | |
Baculovirus | |
Recombinant, Catalytic | |
Serine/Threonine Kinases |
Kinase Assay | |
100 μg | |
STKR Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton™ X-100, 2 mM DTT, and 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |