Learn More
Macherey-Nagel™ Acylating HFBA Pack
Brand: Macherey-Nagel™ 701110.110
Specifications
10 mL | |
1 mL | |
Acylating HFBA Pk. |
Safety and Handling
- HFBA, 1x10 mL
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Heptafluorobutyric anhydride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.