Learn More
KD Scientific Adagio Syringe Pump Control Software
For Legato™ Syringe Pumps. One license for software on USB key.
Brand:  KD Scientific 78-8000
Product Code. 13433649
  • £621.00 / Each
Explore more special offers
Description
Description
  • Low cost simple installation
  • Flow evolution graph
  • Import and Export programs
  • Quick and easy manual pump control
  • Monitor one or more pumps
  • Program data logging
Specifications
Specifications
Control Software
0/1 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
KD Scientific Adagio Syringe Pump Control Software >