missing translation for 'onlineSavingsMsg'
Learn More

Adamantane-1-carbohydrazide, 97%, Thermo Scientific™

£58.70 - £117.00

Chemical Identifiers

CAS 17846-15-0
Molecular Formula C11H18N2O
Molecular Weight (g/mol) 194.278
InChI Key FJXQVRADKYKERU-UHFFFAOYSA-N
Synonym adamantanecarbohydrazide, 1-adamantanecarbohydrazide, adamantane-1-carboxylic acid hydrazide, tricyclo 3.3.1.1∼3,7∼ decane-1-carbohydrazide, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, hydrazide, pubchem21688, hydrazino 1-adamantyl ketone, 3r,5r,7r-adamantane-1-carbohydrazide, z-adamantane-1-carbohydrazonic acid, 3r,5s,7s-adamantane-1-carbohydrazide
PubChem CID 337203
IUPAC Name adamantane-1-carbohydrazide
SMILES C1C2CC3CC1CC(C2)(C3)C(=O)NN
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10160842
View Documents
Thermo Scientific Maybridge
RH01025DA
1 g Amber Glass Bottle
£58.70
1g
Please to purchase this item. Need a web account? Register with us today!
10275884
View Documents
Thermo Scientific Maybridge
RH01025DE
5 g Amber Glass Bottle
£117.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

17846-15-0
194.278
adamantanecarbohydrazide, 1-adamantanecarbohydrazide, adamantane-1-carboxylic acid hydrazide, tricyclo 3.3.1.1∼3,7∼ decane-1-carbohydrazide, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, hydrazide, pubchem21688, hydrazino 1-adamantyl ketone, 3r,5r,7r-adamantane-1-carbohydrazide, z-adamantane-1-carbohydrazonic acid, 3r,5s,7s-adamantane-1-carbohydrazide
adamantane-1-carbohydrazide
C11H18N2O
FJXQVRADKYKERU-UHFFFAOYSA-N
337203
C1C2CC3CC1CC(C2)(C3)C(=O)NN

Specifications

17846-15-0
C11H18N2O
FJXQVRADKYKERU-UHFFFAOYSA-N
adamantane-1-carbohydrazide
337203
97%
97%
adamantanecarbohydrazide, 1-adamantanecarbohydrazide, adamantane-1-carboxylic acid hydrazide, tricyclo 3.3.1.1∼3,7∼ decane-1-carbohydrazide, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, hydrazide, pubchem21688, hydrazino 1-adamantyl ketone, 3r,5r,7r-adamantane-1-carbohydrazide, z-adamantane-1-carbohydrazonic acid, 3r,5s,7s-adamantane-1-carbohydrazide
C1C2CC3CC1CC(C2)(C3)C(=O)NN
194.278
194.28
Adamantane-1-carbohydrazide
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.