missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Adapt-a-Rack™

Product Code. 15340370 Shop All Fisherbrand Products
Click to view available options
Color:
Blue/Blue
Green/Green
Pink/Pink
White/White
Yellow/Yellow
Unit Size:
Pack of 2

Product Code. 15340370

Only null left
Add to basket
Add to basket

Accommodates tubes 12 to 30 mm dia. (5 to 50 mL). Adapt-a-Rack™ holds tubes in a firm, upright position.

  • Made from Polyoxymethylene (POM).
  • Racks can be linked together for additional holding capacity (Linked racks should be supported underneath when carrying).
  • Open sides allow for label viewing.
  • Flat base to hold tubes upright.
  • Available in solid or a combination of colors for easy coding identification.
  • Autoclavable.

Specifications

Product Type Multi Stand Tube Rack
Array 1 x 4
Autoclavable Autoclavable
Color Blue/Blue
Holds Tubes (5 to 50 mL)
Material POM
Positions 4
Shape Rectangular, Rounded Corners
Shelves None
Stackable Yes, Interlocking
Sterility Non-sterile
Quantity 2 Pk.
Width (Metric) 5.6 cm
Height (Metric) 7.6 cm
Diameter (Metric) Well 12 to 30 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.