Learn More
Eppendorf™ Adapter, for 12 tubes 30 mL, for Rotor S-4xUniversal-Large, 2 pcs.
Brand: Eppendorf™ 5920743007
This item is not returnable.
View return policy
Description
Adapter, for 12 tubes 30 mL, for Rotor S-4xUniversal-Large, 2 pcs.Specifications
Adapters, Buckets, and Accessories | |
Rotor S-4 xUniversal-Large | |
12 | |
2 |