Learn More
Eppendorf™ Adapter for Rotor F-35-6-30
Adapter for Rotors designed for use with Eppendorf Centrifuges
£50.30 - £138.00
Product Code | Brand | Capacity | Bore Size (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Bore Size (Metric) | Price | Quantity & Availability | ||||
10379873
|
Eppendorf™
5427735003 |
1 x 9 to 15 mL Tube | Small rotor bore |
£93.15
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10259013
|
Eppendorf™
5427738002 |
1 x 9 to 15 mL Tube | Large rotor bore |
£93.15
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10460184
|
Eppendorf™
5427726004 |
1 x 15 mL Tube | Small rotor bore |
£84.95
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10676654
|
Eppendorf™
5427736000 |
1 x 30 mL Tube | Large rotor bore |
£75.40
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10541944
|
Eppendorf™
5427727000 |
1 x 50 mL Conical Tube | Large rotor bore |
£50.30
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10628004
|
Eppendorf™
5427731008 |
1 x 2.6 to 7 mL Tube | Small rotor bore |
£93.15
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10321254
|
Eppendorf™
5427723005 |
1 x 100 to 125 mm Centriprep™ | Large rotor bore |
£138.00
Pack of 6 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10638774
|
Eppendorf™
5427737006 |
6 x 50 mL | Large rotor bore |
£75.40
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10319833
|
Eppendorf™
5427733000 |
1 x 2.6 to 7 mL Tube | Large rotor bore |
£93.15
Pack of 2 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Rotor F-35-6-30 holds 6 x 15/50mL conical or 12 × Vacutainer™ tubes
Specifications
Rotor F-35-6-30 |