Learn More
BRAND™ Polypropylene Adapter for Pipette Tips
Polypropylene construction. For use with 100μL pipette tips.
Brand:  BRAND™ 25961
8.93 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 10653601
  • £9.40 / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polypropylene construction
  • For 100μL pipette tips
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
Tip Adapter
100μL pipette tips
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ Polypropylene Adapter for Pipette Tips > For use with 100μL pipette tips