Learn More
SCHOTT AG Lighting and Imaging™ Adapter Set for VisiLED Series Illuminators
Combi-kit, adapter set for objective working distances 30 to 110 mm.
Brand: SCHOTT AG Lighting and Imaging™
Code : A0
Additional Details : Weight : 0.48000kg
Description
- Adapter Set to mount VisiLed Darkfield Ring Light (400 300) to VisiLed Brightfield Ring Light (400255, 400225 or 400 100).
Specifications
Adapter Kit | |
Brightfield, Darkfield Adapter Kit | |
Stereo Microscopy Illumination | |
VisiLED Darkfield and Brightfield Ringlight |