Learn More
Eppendorf™ Adapters for 1.5/2.0mL Microcentrifuge Rotors
Use these adaptors with a variety of tubes and rotors. Eppendorf™ Adapters work with all 1.5/2.0mL microcentrifuge rotors.
Brand:  Eppendorf™ 5425723008
Code : 99
Additional Details : Weight : 0.00500kg
 View more versions of this product
Product Code. 10227810
  • £15.75 / Pack of 6
In Stock
to see stock.
Description
Description
  • Adaptors for rotors
  • Available in packs of 6
Specifications
Specifications
Adapter
6 x 0.25 to 0.4 mL Microtainer Tubes
0.2mL PCR Tubes
PCR
Rotor F-45-72-8
6/Pk.